Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

147710

Sigma-Aldrich

Diethyl cyclopropane-1,1-dicarboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H4(CO2C2H5)2
CAS Number:
Molecular Weight:
186.21
Beilstein:
2049509
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.433 (lit.)

bp

94-96 °C/10 mmHg (lit.)

density

1.055 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)C1(CC1)C(=O)OCC

InChI

1S/C9H14O4/c1-3-12-7(10)9(5-6-9)8(11)13-4-2/h3-6H2,1-2H3

InChI key

KYYUCZOHNYSLFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl cyclopropane-1,1-dicarboxylate participates in ring-opening addition reactions with various nucleophilic reagents[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nucleophilic ring-opening additions to 1, 1-disubstituted cyclopropanes.
Stewart JM and Westberg HH.
The Journal of Organic Chemistry, 30(6), 1951-1955 (1965)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service