Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14766

Sigma-Aldrich

2-[2-(Boc-amino)ethoxy]ethoxyacetic acid (dicyclohexylammonium) salt

≥98.0% (TLC)

Synonym(s):

Boc-8-amino-3,6-dioxaoctanoic acid (dicyclohexylammonium) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21NO6 · C12H23N
CAS Number:
Molecular Weight:
444.61
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0% (TLC)

reaction suitability

reaction type: Pegylations
reagent type: cross-linking reagent

SMILES string

C1CCC(CC1)NC2CCCCC2.CC(C)(C)OC(=O)NCCOCCOCC(O)=O

InChI

1S/C12H23N.C11H21NO6/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-11(2,3)18-10(15)12-4-5-16-6-7-17-8-9(13)14/h11-13H,1-10H2;4-8H2,1-3H3,(H,12,15)(H,13,14)

InChI key

SWUXEYKTUQROOO-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polyethylene glycol (PEG) reagents offer favorable characteristics for various applications, including biocompatibility and solubility.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service