Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

147591

Sigma-Aldrich

2-Nitro-1-propanol

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(NO2)CH2OH
CAS Number:
Molecular Weight:
105.09
Beilstein:
1748592
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

liquid

refractive index

n20/D 1.439 (lit.)

bp

72-74 °C/1 mmHg (lit.)

density

1.185 g/mL at 25 °C (lit.)

SMILES string

CC(CO)[N+]([O-])=O

InChI

1S/C3H7NO3/c1-3(2-5)4(6)7/h3,5H,2H2,1H3

InChI key

PCNWBUOSTLGPMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Nitro-1-propanol inhibits methane production during in vitro ruminal fermentation[1]. It is a useful antimicrobial supplement which reduces carriage of certain food-borne pathogens in animals[2].

Application

2-Nitro-1-propanol was used in improved synthesis of 2,2,2-trinitroethyl ethers[3].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic Communications, 20, 3303-3303 (1990)
Y S Jung et al.
Letters in applied microbiology, 39(5), 471-476 (2004-10-16)
To test the inhibitory activity of 2-nitro-1-propanol (2NPOH) against Salmonella Typhimurium, Escherichia coli O157:H7 and Enterococcus faecalis. Specific growth rates (h(-1)) of S. Typhimurium, E. coli O157:H7 and Ent. faecalis were determined during culture in tryptic soya broth (TSB) supplemented
Robin C Anderson et al.
Bioresource technology, 99(18), 8655-8661 (2008-06-10)
Ruminal methane (CH(4)) production results in the loss of up to 12% of gross energy intake and contributes nearly 20% of the United States' annual emission of this greenhouse gas. We report the effects of select nitrocompounds on ruminal fermentation
M Dimitrijevic et al.
Journal of food protection, 69(5), 1061-1065 (2006-05-24)
We report the effects of 2-nitro-1-propanol (2NPOH), 2-nitroethanol (2NEOH), and nitroethane (NE) on growth and survivability of Listeria monocytogenes. In all cases, inhibition was greatest with 2NPOH and least with NE. For example, specific growth rates of L. monocytogenes strain
L A Donaldson et al.
Journal of microscopy, 259(3), 228-236 (2015-05-01)
A novel approach to nanoscale detection of cell wall porosity using confocal fluorescence microscopy is described. Infiltration of cell walls with a range of nitrophenyl-substituted carbohydrates of different molecular weights was assessed by measuring changes in the intensity of lignin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service