Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14735

Sigma-Aldrich

N,N′-Bis(2-dimethylaminoethyl)-N,N′-dimethyl-9,10-anthracenedimethanamine

≥97.0% (CHN)

Synonym(s):

9,10-Bis{N-[2-(dimethylamino)ethyl]methylaminomethyl}anthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H38N4
CAS Number:
Molecular Weight:
406.61
Beilstein:
3628941
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (CHN)

SMILES string

CN(C)CCN(C)Cc1c2ccccc2c(CN(C)CCN(C)C)c3ccccc13

InChI

1S/C26H38N4/c1-27(2)15-17-29(5)19-25-21-11-7-9-13-23(21)26(20-30(6)18-16-28(3)4)24-14-10-8-12-22(24)25/h7-14H,15-20H2,1-6H3

InChI key

KOLPZKQPBGVSAU-UHFFFAOYSA-N

Application

N,N′-Bis(2-dimethylaminoethyl)-N,N′-dimethyl-9,10-anthracenedimethanamine was used as ionophore in liquid membrane based Zn2+ ion selective electrode.

Other Notes

Chelation enhanced fluorescence with zinc ions

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potentiometric studies of N, N′-Bis (2-dimethylaminoethyl)- N, N′-dimethyl-9, 10 anthracenedimethanamine as a chemical sensing material for Zn (II) ions.
Gupta NR, et al.
Materials Science & Engineering. C, Materials For Biological Applications, 28(7), 1025-1030 (2008)
M.E. Huston et al.
Journal of the American Chemical Society, 110, 4460-4460 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service