Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

145793

Sigma-Aldrich

4-Chloro-2-methoxybenzoic acid

99%

Synonym(s):

4-Chloro-o-anisic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(OCH3)CO2H
CAS Number:
Molecular Weight:
186.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

powder

mp

146-148 °C (lit.)

functional group

carboxylic acid
chloro

SMILES string

COc1cc(Cl)ccc1C(O)=O

InChI

1S/C8H7ClO3/c1-12-7-4-5(9)2-3-6(7)8(10)11/h2-4H,1H3,(H,10,11)

InChI key

UFEYMXHWIHFRBX-UHFFFAOYSA-N

General description

4-Chloro-2-methoxybenzoic acid anion forms complexes with Mn2+, Co2+, Ni2+, Cu2+ and Zn2+[1].

Application

4-Chloro-2-methoxybenzoic acid was used in the synthesis of (2-(4-chloro-2-methoxybenzoyl)-1′H-1,3′-bipyrrol-2′-yl)(2-methoxyphenyl)methanone[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Liu et al.
Marine drugs, 10(4), 953-962 (2012-06-13)
Infections caused by drug-resistant pathogens are on the rise. The ongoing spread of methicillin-resistant Staphylococcus aureus (MRSA) strains exemplifies the urgent need for new antibiotics. The marine natural product, marinopyrrole A, was previously shown to have potent antibiotic activity against
Complexes of Mn (II), Co (II), Ni (II), Cu (II) and Zn (II) with 4-chloro-2-methoxybenzoic acid anion.
Ferenc, et al.
Journal of Thermal Analysis and Calorimetry, 86(3), 783-789 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service