Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

145769

Sigma-Aldrich

4-Bromo-α-methylbenzyl alcohol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH(CH3)OH
CAS Number:
Molecular Weight:
201.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

bp

119-121 °C/7 mmHg (lit.)

mp

36-38 °C (lit.)

density

1.46 g/mL at 25 °C (lit.)

functional group

bromo
hydroxyl

SMILES string

CC(O)c1ccc(Br)cc1

InChI

1S/C8H9BrO/c1-6(10)7-2-4-8(9)5-3-7/h2-6,10H,1H3

InChI key

XTDTYSBVMBQIBT-UHFFFAOYSA-N

Application

4-Bromo-α-methylbenzyl alcohol was used in the synthesis of 3-(4-(1-(tert-butyl)dimethylsilyloxy)ethyl)phenyl)-3-(trifluoromethyl)-3H-diazirine and (1-(4-bromophenyl)ethoxy)(tert-butyl)dimethylsilane.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Shaukat Husain et al.
Journal of medicinal chemistry, 53(17), 6432-6444 (2010-08-14)
We synthesized the R- and S-enantiomers of ethyl 1-(1-(4-(3-((trifluoromethyl)-3H-diazirin-3-yl)phenyl)ethyl)-1H-imidazole-5-carboxylate (trifluoromethyldiazirinyl-etomidate), or TFD-etomidate, a novel photoactivable derivative of the stereoselective general anesthetic etomidate (R-(2-ethyl 1-(phenylethyl)-1H-imidazole-5-carboxylate)). Anesthetic potency was similar to etomidate's, but stereoselectivity was reversed and attenuated. Relative to etomidate, TFD-etomidate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service