Skip to Content
MilliporeSigma
All Photos(1)

Documents

144045

Sigma-Aldrich

Diethyl tetradecanedioate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5O2C(CH2)12CO2C2H5
CAS Number:
Molecular Weight:
314.46
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

solid

mp

29-31 °C (lit.)

SMILES string

CCOC(=O)CCCCCCCCCCCCC(=O)OCC

InChI

1S/C18H34O4/c1-3-21-17(19)15-13-11-9-7-5-6-8-10-12-14-16-18(20)22-4-2/h3-16H2,1-2H3

InChI key

XXYWYFSQJPFXBD-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service