Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

144029

Sigma-Aldrich

2,6-Dimethylquinoline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11N
CAS Number:
Molecular Weight:
157.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 144029, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

98%

form

solid

mp

57-59 °C (lit.)

SMILES string

Cc1ccc2nc(C)ccc2c1

InChI

1S/C11H11N/c1-8-3-6-11-10(7-8)5-4-9(2)12-11/h3-7H,1-2H3

InChI key

JJPSZKIOGBRMHK-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Application

2,6-Dimethylquinoline was used to study the inhibition potencies (IC50 values) of structurally diverse chemicals with recombinant human CYP2B6 enzyme for in vitro research purposes[1].

Biochem/physiol Actions

2,6-Dimethylquinoline is the chemical constituent present in roots of Peucedantu praeruptorum[2]. It is a potential inhibitor of cytochrome P450 1A2 activity[3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L E Korhonen et al.
British journal of pharmacology, 150(7), 932-942 (2007-02-28)
The cytochrome P450 2B6 (CYP2B6) enzyme metabolises a number of clinically important drugs. Drug-drug interactions resulting from inhibition or induction of CYP2B6 activity may cause serious adverse effects. The aims of this study were to construct a three-dimensional structure-activity relationship
Cun Zhang et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 31(16), 1333-1335 (2006-10-26)
To isolate and identify the chemical constituents from the roots of Peucedantu praeruptorum. The constituents were isolated by column chromatography on silica gel and ODS, and identified by NMR, MS spectroscopic methods. Eight compounds, (-) sclerodin (1), palmitic acid (2)
Markus Brinkmann et al.
Chemical research in toxicology, 32(4), 698-707 (2019-03-22)
Hydroxylation of polyaromatic compounds through cytochromes P450 (CYPs) is known to result in potentially estrogenic transformation products. Recently, there has been an increasing awareness of the importance of alternative pathways such as aldehyde oxidases (AOX) or N-methyltransferases (NMT) in bioactivation
Bin Yang et al.
Environmental science and pollution research international, 23(4), 3399-3405 (2015-10-23)
The solubilities of 19 different kinds of N-heteroaromatic compounds in aqueous solutions with different concentrations of NaCl were determined at 298.15 K with a UV-vis spectrophotometry and titration method, respectively. Setschenow constants, Ks, were employed to describe the solubility behavior
Laura E Korhonen et al.
Journal of medicinal chemistry, 48(11), 3808-3815 (2005-05-27)
The purpose of this study was to determine the cytochrome P450 1A2 (CYP1A2) inhibition potencies of structurally diverse compounds to create a comprehensive three-dimensional quantitative structure-activity relationship (3D-QSAR) model of CYP1A2 inhibitors and to use this model to predict the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service