Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

143693

Sigma-Aldrich

2-Methoxyethylamine

98%

Synonym(s):

2-Aminoethyl methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2NH2
CAS Number:
Molecular Weight:
75.11
Beilstein:
741854
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.406 (lit.)

bp

95 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

SMILES string

COCCN

InChI

1S/C3H9NO/c1-5-3-2-4/h2-4H2,1H3

InChI key

ASUDFOJKTJLAIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Courtney H Fox et al.
Nature communications, 6, 7417-7417 (2015-07-16)
Dynamic covalent materials are stable materials that possess reversible behaviour triggered by stimuli such as light, redox conditions or temperature; whereas supramolecular crosslinks depend on the equilibrium constant and relative concentrations of crosslinks as a function of temperature. The combination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service