Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

142956

Sigma-Aldrich

2,4-Dinitrobenzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3SO2Cl
CAS Number:
Molecular Weight:
266.62
Beilstein:
2147583
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

101-103 °C (lit.)

functional group

nitro

SMILES string

[O-][N+](=O)c1ccc(c(c1)[N+]([O-])=O)S(Cl)(=O)=O

InChI

1S/C6H3ClN2O6S/c7-16(14,15)6-2-1-4(8(10)11)3-5(6)9(12)13/h1-3H

InChI key

SSFSNKZUKDBPIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dinitrobenzenesulfonyl chloride causes the sulfonation of glycosylamines to yield N-glycosyl-2,4-dinitrobenzenesulfonamides.
May contain up to 3.5% benzene

Application

2,4-Dinitrobenzenesulfonyl chloride was used to protect primary amines. It was used as starting reagent in the synthesis of tert-butyl 2-[(2,4-dinitrophenyl) sulfonyl]aminoacetate.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A - Eye Dam. 1 - Muta. 1B - Skin Corr. 1B - STOT RE 2

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachel J Ball et al.
Artificial DNA, PNA & XNA, 1(1), 27-35 (2011-06-21)
Halogen-labelled peptide organic acid (HPOA) monomers have been synthesised and incorporated into sequence-specific peptide nucleic acid (PNA) probes. Three different types of probe have been prepared; the unmodified PNA probe, the PNA probe with a mass marker, and the PNA
Vishwanath Gaitonde et al.
Journal of carbohydrate chemistry, 31(4-6), 353-370 (2013-01-26)
The N-glycosyl-2,4-dinitrobenzenesulfonamides were accessed via benzoyl-protected β-glycosyl azides. The azides were reduced with Adams' catalyst to the corresponding amines. The glycosylamines were sulfonated with 2,4-dinitrobenzenesulfonyl chloride to form N-glycosyl-2,4-dinitrobenzenesulfonamides in moderate yields. β-Glycosyl amides were then prepared in 67 -

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service