Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

141666

Sigma-Aldrich

1-(2-Aminoethyl)piperidine

98%

Synonym(s):

2-Piperidinoethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16N2
CAS Number:
Molecular Weight:
128.22
Beilstein:
103469
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.473 (lit.)

bp

186 °C (lit.)

density

0.899 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

NCCN1CCCCC1

InChI

1S/C7H16N2/c8-4-7-9-5-2-1-3-6-9/h1-8H2

InChI key

CJNRGSHEMCMUOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(2-Aminoethyl)piperidine on [1+1] condensation reaction with 3-methoxy salicylaldehyde yields tridentate Schiff base ligand[1].

Application

1-(2-Aminoethyl)piperidine was used in modification of vinylbenzyl chloride/divinylbenzene gel copolymer bead[2]. It was used in the synthesis of linkage isomers trans-bis[1-(2-aminoethyl)piperidine]dinitronickel and trans-bis[1-(2-aminoethyl)-piperidine]dinitritonickel[3].
Reactant for synthesis of:
Analogs of anticancer agents
Inhibitor of Botulinum Neurotoxin Serotype A light Chain, P. falciparum Malaria, and Ebola Filovirus
Cannabinoid CB1 receptor antagonists
Small molecules that restore E-cadherin expression and reduve invasion in colorectal carcinoma cells
Potent and selective 5-HT6 antagonists
N-mustards as anticancer agents

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

First structural characterisation of nitro-nitrito linkage isomers of nickel (II): synthesis and single crystal structures of [NiL2 (NO2) 2] and [NiL2 (ONO) 2][La= a1-(2-aminoethyl) piperidine].
Das D, et al.
J. Chem. Soc., Dalton Trans., 23, 3987-3990 (1998)
Sujit Sasmal et al.
Inorganic chemistry, 50(12), 5687-5695 (2011-05-31)
The work in this paper presents the syntheses, structures, and magnetic properties of three end-to-end (EE) azide/cyanate-bridged copper(II) compounds [Cu(II)L(1)(μ(1,3)-NCO)](n)·2nH(2)O (1), [Cu(II)L(1)(μ(1,3)-N(3))](n)·2nH(2)O (2), and [Cu(II)L(2)(μ(1,3)-N(3))](n) (3), where the ligands used to achieve these species, HL(1) and HL(2), are the tridentate
Dorota Jermakowicz-Bartkowiak
Environmental geochemistry and health, 32(4), 317-320 (2010-04-14)
Vinylbenzyl chloride/divinylbenzene gel copolymer beads have been modified using piperidine, hexamethyleneimine, piperazine, 1-(2-aminoethyl)piperazine, and N-cyclohexyl-1,3-propanediamine. These resins were then tested for the sorption of noble metal ions, namely, Re(VII), Pd(II), Pt(IV), and Au(III), from a 0.1 M HCl solution. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service