Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

141216

Sigma-Aldrich

(3,4-Dimethoxyphenyl)acetone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2COCH3
CAS Number:
Molecular Weight:
194.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.5358 (lit.)

density

1.115 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(CC(C)=O)cc1OC

InChI

1S/C11H14O3/c1-8(12)6-9-4-5-10(13-2)11(7-9)14-3/h4-5,7H,6H2,1-3H3

InChI key

UMYZWICEDUEWIM-UHFFFAOYSA-N

General description

3,4-Dimethoxyphenylacetone undergoes asymmetric amination catalyzed by Brevibacterium linens IFO 12141 strain to yield corresponding optically active amine[1].

Application

3,4-Dimethoxyphenylacetone was used as starting material in the synthesis of α-methyl-dopa, an important nonproteogenic α-amino acid for pharmaceutical applications[2].

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric amination of 4-methoxyphenylacetone and its related compounds with microorganisms.
Nakamichi K, et al.
Applied Microbiology and Biotechnology, 33(6), 637-640 (1990)
W H Boesten et al.
Organic letters, 3(8), 1121-1124 (2001-05-12)
[reaction: see text]. Diastereoselective Strecker reactions based on (R)-phenylglycine amide as chiral auxiliary are reported. The Strecker reaction is accompanied by an in situ crystallization-induced asymmetric transformation, whereby one diastereomer selectively precipitates and can be isolated in 76-93% yield and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service