Skip to Content
MilliporeSigma
All Photos(2)

Documents

140996

Sigma-Aldrich

Ethyl crotonate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCOOC2H5
Molecular Weight:
114.14
Beilstein:
635834
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.9 (vs air)

Assay

96%

form

liquid

refractive index

n20/D 1.424 (lit.)

bp

142-143 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)\C=C\C

InChI

1S/C6H10O2/c1-3-5-6(7)8-4-2/h3,5H,4H2,1-2H3/b5-3+

InChI key

ZFDIRQKJPRINOQ-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl crotonate on hydroboration yields stereospecifically 2-tritiated 3-hydroxybutyric acid.

Application

Ethyl crotonate was used in chemical-shift imaging during the rapid analysis of multiple samples using multiplex sample NMR. It was used as reagent during the total synthesis of (+/-)-daurichromenic acid.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Willadsen et al.
European journal of biochemistry, 54(1), 247-252 (1975-05-01)
1. A specimen of stereospecifically 2-tritiated 3-hydroxybutyric acid was prepared by hydroboration of ethyl crotonate. It was assumed that the hydroboration reaction took a syn course and hence that (2R,3S) plus (2S,3R)-3-hydroxy[2 minus 3H1]butyric acid was formed after oxidation and
Hongjuan Hu et al.
The Journal of organic chemistry, 69(11), 3782-3786 (2004-05-22)
A modular and concise total synthesis of (+/-)-daurichromenic acid has been accomplished in four steps from ethyl acetoacetate, ethyl crotonate, and trans,trans-farnesal. A series of analogues of this natural product, which has potent anti-HIV activity, were also prepared from ethyl
T Hou et al.
Analytical chemistry, 73(11), 2541-2546 (2001-06-14)
Two improved approaches for the rapid analysis of multiple samples using multiplex sample NMR are described. In the first approach, frequency-selective 90 degrees radio frequency pulses and large pulsed field gradients are applied to excite and detect multiple samples in
Amélie Slegers et al.
Molecules (Basel, Switzerland), 20(6), 10980-11016 (2015-06-18)
Developed from crosses between Vitis vinifera and North American Vitis species, interspecific hybrid grape varieties are becoming economically significant in northern areas, where they are now extensively grown for wine production. However, the varietal differences between interspecific hybrids are not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service