Skip to Content
MilliporeSigma
All Photos(1)

Documents

140562

Sigma-Aldrich

5-Chloroisatin

97%

Synonym(s):

5-Chloro-1H-indole-2,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4ClNO2
CAS Number:
Molecular Weight:
181.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

254-258 °C (lit.)

SMILES string

Clc1ccc2NC(=O)C(=O)c2c1

InChI

1S/C8H4ClNO2/c9-4-1-2-6-5(3-4)7(11)8(12)10-6/h1-3H,(H,10,11,12)

InChI key

XHDJYQWGFIBCEP-UHFFFAOYSA-N

General description

5-Chloroisatin reacts with substituted 4-amino-4,5-dihydro-1H-1,2,4-triazole-5-ones to yield Schiff bases.

Application

5-Chloroisatin was used in synthesis of Schiff base by reaction with 2-methyl-4-nitroaniline.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Olcay Bekircan et al.
Molecules (Basel, Switzerland), 13(9), 2126-2135 (2008-10-03)
Ethyl imidate hydrochlorides 1 were prepared by passing HCl gas through solutions of substituted benzyl cyanides and absolute ethanol. Ethoxycarbonylhydrazones 2 were synthesized from the reaction of compounds 1 with ethyl carbazate. Treatment of 2 with hydrazine hydrate leads to
Synthesis, Characterization and Biological Activities of a New 5-Chloroisatin Schiff Base and its Metal Complexes.
RAO R, et al.
Chemical Science Transactions, 2(3), 1063-1069 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service