Skip to Content
MilliporeSigma
All Photos(1)

Documents

14010

Sigma-Aldrich

O-Benzyl-L-tyrosine

≥99.0% (NT), for peptide synthesis

Synonym(s):

4-Benzyloxy-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
271.31
Beilstein:
2659642
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

product name

O-Benzyl-L-tyrosine, ≥99.0% (NT)

Assay

≥99.0% (NT)

optical activity

[α]20/D −9.5±1°, c = 1% in acetic acid: water (4:1)

reaction suitability

reaction type: solution phase peptide synthesis

mp

259 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

N[C@@H](Cc1ccc(OCc2ccccc2)cc1)C(O)=O

InChI

1S/C16H17NO3/c17-15(16(18)19)10-12-6-8-14(9-7-12)20-11-13-4-2-1-3-5-13/h1-9,15H,10-11,17H2,(H,18,19)/t15-/m0/s1

InChI key

KAFHLONDOVSENM-HNNXBMFYSA-N

Gene Information

human ... PPARG(5468)

Looking for similar products? Visit Product Comparison Guide

General description

O-Benzyl-L-tyrosine is a derivative of the natural amino acid tyrosine. It is used as a building block in a variety of applications, including solution-phase peptide synthesis, and the production of pharmaceuticals and other chemicals.

Application

O-Benzyl-L-tyrosine can be used as a building block in the synthesis of amphiphilic block co-polypeptides, due to their potential applications in biomedical fields. For instance, it is used to prepare Lysine-block-tyrosine block copolypeptides, which are useful as targeted drug carriers, functional nanobioreactors, and biomimetic encapsulants in the biomedical fields.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service