Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

14001

Sigma-Aldrich

Benzyltrimethylammonium tetrachloroiodate

≥98.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(ICl4)(CH3)3
CAS Number:
Molecular Weight:
418.96
Beilstein:
4619219
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (AT)

form

solid

SMILES string

Cl[I-](Cl)(Cl)Cl.C[N+](C)(C)Cc1ccccc1

InChI

1S/C10H16N.Cl4I/c1-11(2,3)9-10-7-5-4-6-8-10;1-5(2,3)4/h4-8H,9H2,1-3H3;/q+1;-1

InChI key

IBXYFQYYVRYALP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Stable, selective and crystalline chlorinating agent for various compounds

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bulletin of the Chemical Society of Japan, 63, 941-941 (1990)
Bulletin of the Chemical Society of Japan, 62, 2096-2096 (1989)
Chemistry Letters (Jpn), 415-415 (1989)
Alicia Dubinski et al.
Human molecular genetics, 32(2), 319-332 (2022-08-23)
Responding effectively to external stress is crucial for neurons. Defective stress granule dynamics has been hypothesized as one of the pathways that renders motor neurons in amyotrophic lateral sclerosis (ALS) more prone to early death. Specifically, it is thought that
S. Kajigaeshi et al.
Tetrahedron Letters, 29, 5783-5783 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service