Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

139076

Sigma-Aldrich

5-Bromo-4-chloroindoxyl 1,3-diacetate

98%

Synonym(s):

5-Bromo-4-chloro-3-indolyl 1,3-diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9BrClNO3
CAS Number:
Molecular Weight:
330.56
Beilstein:
270885
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

165-168 °C (lit.)

solubility

chloroform: soluble 50 mg/mL, clear, pale yellow to yellow

storage temp.

2-8°C

SMILES string

CC(=O)Oc1cn(C(C)=O)c2ccc(Br)c(Cl)c12

InChI

1S/C12H9BrClNO3/c1-6(16)15-5-10(18-7(2)17)11-9(15)4-3-8(13)12(11)14/h3-5H,1-2H3

InChI key

DSHQTSIXXYZXGR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Bromo-4-chloroindoxyl 1,3-diacetate was used as starting reagent in the synthesis of 5,5′-dibromo-4,4′-dichloroindigo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Faten Arab-Jaziri et al.
New biotechnology, 30(5), 536-544 (2013-05-01)
Directed evolution was applied to the α-l-arabinofuranosidase from Thermobacillus xylanilyticus to confer better transglycosylation ability, particularly for the synthesis of benzyl α-l-arabinofuranosyl-(1,2)-α-d-xylopyranoside, starting from p-nitrophenyl α-l-arabinofuranoside (donor) and benzyl α-d-xylopyranoside (acceptor). The aim was to obtain mutants displaying both lower

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service