Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

138940

Sigma-Aldrich

3,4,5-Trichloropyridazine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4HCl3N2
CAS Number:
Molecular Weight:
183.42
Beilstein:
127119
EC Number:
MDL number:

Assay

98%

SMILES string

Clc1cnnc(Cl)c1Cl

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J J Freeman et al.
Contact dermatitis, 18(5), 259-262 (1988-05-01)
The potential of 3,4,5-trichloropyridazine to induce contact sensitization was assessed in the guinea pig maximization test of Magnusson and Kligman and also in the closed patch test described by Buehler. The test material was a 1% solution of 3,4,5-trichloropyridazine in
T G Warshaw
Contact dermatitis, 18(5), 257-258 (1988-05-01)
A primary irritant dermatitis to 3,4,5-trichloropyridazine was observed on the hands of a chemist after accidental exposure. Healing was rapid in comparison to a report on 3,4,6-trichloropyridazine toxicity. Prompt treatment with a potent topical corticosteroid, wet soaks, and an oral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service