Skip to Content
MilliporeSigma
All Photos(1)

Documents

138894

Sigma-Aldrich

2,4,5-Trimethoxybenzoic acid

99%

Synonym(s):

Asaronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CO2H
CAS Number:
Molecular Weight:
212.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

143-145 °C (lit.)

SMILES string

COc1cc(OC)c(cc1OC)C(O)=O

InChI

1S/C10H12O5/c1-13-7-5-9(15-3)8(14-2)4-6(7)10(11)12/h4-5H,1-3H3,(H,11,12)

InChI key

KVZUCOGWKYOPID-UHFFFAOYSA-N

Application

2,4,5-Trimethoxybenzoic acid was used as internal standard during the determination of the various types of hydroxyl groups present in lignins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative phosphorus-31 NMR analysis of lignins, a new tool for the lignin chemist.
Argyropoulos DS.
J. Wood Chem. Technol., 14(1), 45-63 (1994)
A new synthesis of asaronic acid and aldehyde and their derivatives.
Rajagopalan S, et al.
Proceedings of the Indian Academy of Sciences - Section A, 30(5), 265-270 (1949)
Hyeongjoo Oh et al.
Nutrients, 12(7) (2020-07-10)
Macrophage polarization has been implicated in the pathogenesis of metabolic diseases such as obesity, diabetes, and atherosclerosis. Macrophages responsiveness to polarizing signals can result in their functional phenotype shifts. This study examined whether high glucose induced the functional transition of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service