Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

138177

Sigma-Aldrich

2,5-Dimethylbenzoic acid

95%

Synonym(s):

p-Xylylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3CO2H
CAS Number:
Molecular Weight:
150.17
Beilstein:
971588
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

bp

268 °C (lit.)

mp

132-134 °C (lit.)

SMILES string

Cc1ccc(C)c(c1)C(O)=O

InChI

1S/C9H10O2/c1-6-3-4-7(2)8(5-6)9(10)11/h3-5H,1-2H3,(H,10,11)

InChI key

XZRHNAFEYMSXRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,5-Dimethylbenzoic acid is a metabolite of pseudocumene (1,2,4-trimethylbenzene).

Application

2,5-Dimethylbenzoic acid was used in the synthesis of new derivatives of [24]paracyclophane. It was used as starting reagent in the synthesis of methyl 2,5-dimethylbenzoate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Substituted [24] paracyclophanes.
Shabtai E , et al.
Journal of the Chemical Society. Perkin Transactions 1, 2(11), 2411-2414 (1997)
Synthesis and properties of photoluminescent polymers bearing electron-facilitating oxadiazole derivative side groups.
Kim JJ, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 40(8), 1173-1183 (2002)
Radosław Swiercz et al.
International journal of occupational medicine and environmental health, 15(1), 37-42 (2002-06-01)
The objective of this study was to evaluate the toxicokinetics and metabolism of pseudocumene after inhalation exposure. Male Wistar rats were exposed to pseudocumene vapors at nominal concentrations of 25,100 or 250 ppm in the dynamic inhalation chambers for 6

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service