Skip to Content
MilliporeSigma
All Photos(1)

Documents

137685

Sigma-Aldrich

2-Bromoethyl acetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2Br
CAS Number:
Molecular Weight:
167.00
Beilstein:
1743110
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

impurities

<3% acetic anhydride

refractive index

n20/D 1.455 (lit.)

bp

159 °C (lit.)

mp

−14-−13.8 °C (lit.)

density

1.514 g/mL at 25 °C (lit.)

SMILES string

CC(=O)OCCBr

InChI

1S/C4H7BrO2/c1-4(6)7-3-2-5/h2-3H2,1H3

InChI key

RGHQKFQZGLKBCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Bromoethyl acetate was used as alkylating reagent during asymmetric total synthesis of (+)-1-deoxylycorine and (+)-lycorine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuya Uto et al.
Physical chemistry chemical physics : PCCP, 20(31), 20669-20677 (2018-07-31)
Chitin is a promising biomass resource and has high potential for industrial applications owing to its huge annual production in nature. However, it exhibits poor processability and solubility due to its very stable and crystalline character. Recently, ionic liquids (ILs)
The first asymmetric total syntheses of (+)-lycorine and (+)-1-deoxylycorine.
Schultz AG, et al.
Journal of the American Chemical Society, 118(26), 6210-6219 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service