Skip to Content
MilliporeSigma
All Photos(1)

Documents

137243

Sigma-Aldrich

4-(p-Tolyl)-3-buten-2-one

(predominantly trans), 97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH=CHCOCH3
CAS Number:
Molecular Weight:
160.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

composition

(predominantly trans)

mp

29-33 °C (lit.)

SMILES string

CC(=O)\C=C\c1ccc(C)cc1

InChI

1S/C11H12O/c1-9-3-6-11(7-4-9)8-5-10(2)12/h3-8H,1-2H3/b8-5+

InChI key

USXMUOFSQBSHGN-VMPITWQZSA-N

General description

4-(p-Tolyl)-3-buten-2-one undergoes Reformatsky reaction with ethyl α-bromopropionate to yield β-hydroxy esters.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geometric Isomers of 2, 3-Dimethyl-5-aryl-2, 4-pentadienoic Acids. The Steric Factors Favoring 2-cis Formation.
Wiley RH, et al.
The Journal of Organic Chemistry, 29(10), 2986-2990 (1964)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service