Skip to Content
MilliporeSigma
All Photos(1)

Documents

136697

Sigma-Aldrich

Hexamethyldewarbenzene

97%

Synonym(s):

1,2,3,4,5,6-Hexamethylbicyclo[2.2.0]hexa-2,5-diene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18
CAS Number:
Molecular Weight:
162.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.448 (lit.)

bp

60 °C/20 mmHg (lit.)

mp

7 °C (lit.)

density

0.803 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1=C(C)C2(C)C(C)=C(C)C12C

InChI

1S/C12H18/c1-7-8(2)12(6)10(4)9(3)11(7,12)5/h1-6H3

InChI key

RVNQQZMIWZPGNA-UHFFFAOYSA-N

General description

Hexamethyl dewar benzene on 1,2-addition with transient W(CO)5-complexed phosphinidenes affords the novel 3-phosphatricyclo[3.2.0.0(2,4)]hept-6-ene complexes. It undergoes partial rearrangement to give hexamethylbenzene during its epoxidation using (tetraphenylporphyrinato)iron(III)chloride and m-chloroperbenzoic acid.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isomerization of hexamethyl (Dewar benzene) to hexamethylbenzene catalyzed by electron acceptors. Thermal generation of an exciplex.
Peacock NJ and Schuster GB.
Journal of the American Chemical Society, 105(11), 3632-3638 (1983)
Erik P A Couzijn et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(5), 1499-1507 (2007-11-23)
1,2-Addition of transient W(CO)(5)-complexed phosphinidenes exo to hexamethyl Dewar benzene affords the novel 3-phosphatricyclo[3.2.0.0(2,4)]hept-6-ene complexes. The fused tricyclic phosphiranes are obtained as both the Z and the thermally less stable E isomers, the (31)P NMR chemical shifts of which differ
Mechanisms of hemin-catalyzed epoxidations: electron transfer from alkenes.
Traylor TG and Miksztal AR.
Journal of the American Chemical Society, 109(9), 2770-2774 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service