Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

136468

Sigma-Aldrich

4′-Piperazinoacetophenone

94%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16N2O
CAS Number:
Molecular Weight:
204.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

94%

form

solid

mp

107-110 °C (lit.)

SMILES string

CC(=O)c1ccc(cc1)N2CCNCC2

InChI

1S/C12H16N2O/c1-10(15)11-2-4-12(5-3-11)14-8-6-13-7-9-14/h2-5,13H,6-9H2,1H3

InChI key

KPXVKKBJROCIJB-UHFFFAOYSA-N

Gene Information

Application

4′-Piperazinoacetophenone was used as sensitizer-linked amine to study the triplet-triplet energy transfer from cation to anion in crystalline salts[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cation to anion triplet-triplet energy transfer in crystalline organic salts.
Gamlin JN, et al.
Tetrahedron Letters, 37(34), 6037-6040 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service