Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13622

Sigma-Aldrich

4-Benzyloxy-2-butanone

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCH2CH2COCH3
CAS Number:
Molecular Weight:
178.23
Beilstein:
779045
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

refractive index

n20/D 1.505

bp

90-92 °C/0.1 mmHg (lit.)

density

1.027 g/mL at 20 °C (lit.)

SMILES string

CC(=O)CCOCc1ccccc1

InChI

1S/C11H14O2/c1-10(12)7-8-13-9-11-5-3-2-4-6-11/h2-6H,7-9H2,1H3

InChI key

BTOZGSIVAZGXCH-UHFFFAOYSA-N

General description

4-Benzyloxy-2-butanone on aminophosphonylation followed by cyclization yields 3-amino-2-oxo-1,2-oxaphospholane[1].

Application

4-Benzyloxy-2-butanone was used in the synthesis of 4-benzyloxy-2-butanol[2].

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 3-amino-2-oxo-1, 2-oxaphospholanes and 3-amino-2-oxo-1, 2-oxaphosphorinanes.
Finet J, et al.
Phosph. Sulfur Relat. Elem., 81(1-4), 17-25 (1993)
Reductive cleavage of acetals and ketals by Borane.
Fleming B and Bolker HI.
Canadian Journal of Chemistry, 52(6), 888-893 (1974)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service