Skip to Content
MilliporeSigma
All Photos(1)

Documents

135542

Sigma-Aldrich

Diethyl benzylmalonate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(CO2C2H5)2
CAS Number:
Molecular Weight:
250.29
Beilstein:
615793
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.486 (lit.)

bp

162-163 °C/10 mmHg (lit.)

density

1.064 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(Cc1ccccc1)C(=O)OCC

InChI

1S/C14H18O4/c1-3-17-13(15)12(14(16)18-4-2)10-11-8-6-5-7-9-11/h5-9,12H,3-4,10H2,1-2H3

InChI key

ICZLTZWATFXDLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl benzylmalonate on condensation with 2-amino-benzimidazole yields 3-benzyl-4-hydroxypyrimido[1,2-a]benzimidazole-2-one.

Application

Diethyl benzylmalonate was used in the synthesis of macrocyclic ligands and determination of stability constants of their Cu(II), Ni(II) and Co(II) complexes. It was used as starting reagent for the synthesis of 2-benzyl-1,3-propane diol.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Tian et al.
Dalton transactions (Cambridge, England : 2003), 47(3), 675-683 (2017-12-23)
A visible light induced three-component catalytic system with the cobalamin derivative (B
Dioxygen adducts of nickel (II) and cobalt (II) dioxopentaazamacrocyclic complexes: Kinetics, stabilities, and hydroxylation of the ligands in the nickel dioxygen complexes.
Chen D, et al.
Inorganic Chemistry, 30(6), 1396-1402 (1991)
Jonathan R LaRochelle et al.
Bioorganic & medicinal chemistry, 25(24), 6479-6485 (2017-11-02)
The PTPN11 oncogene encodes the cytoplasmic protein tyrosine phosphatase SHP2, which, through its role in multiple signaling pathways, promotes the progression of hematological malignancies and other cancers. Here, we employ high-throughput screening to discover a lead chemical scaffold, the benzothiazolopyrimidones
Enzymatic asymmetrization of prochiral 2-benzyl-1, 3-propanediol through esterification in solvent media.
Ducret A, et al.
Biotechnology Letters, 22(8), 709-713 (2000)
A Kreutzberger et al.
Arzneimittel-Forschung, 33(11), 1517-1518 (1983-01-01)
Within the structural class of pyrimidol[1,2-a]benzimidazole-2,4-diones accessible through condensation of 2-amino-benzimidazole (1) with malonates (2) there occur representatives exhibiting centrally dampening properties. In particular, aromatic moieties have now been included into the present investigations. By condensation of 1 with diethyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service