Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

135151

Sigma-Aldrich

3-Chloro-4′-fluoropropiophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4COCH2CH2Cl
CAS Number:
Molecular Weight:
186.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

47-49 °C (lit.)

functional group

chloro
fluoro
ketone

SMILES string

Fc1ccc(cc1)C(=O)CCCl

InChI

1S/C9H8ClFO/c10-6-5-9(12)7-1-3-8(11)4-2-7/h1-4H,5-6H2

InChI key

AAHQPLJUSLMHHR-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isao Kinoyama et al.
Chemical & pharmaceutical bulletin, 52(11), 1330-1333 (2004-11-02)
The search for novel antiandrogens by high-throughput screening (HTS) of the Yamanouchi chemical library led to the discovery of the lead compound (5), which possesses an arylmorpholine moiety. Through the optimization of the lead compound (5), we have found a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service