Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

134155

Sigma-Aldrich

1-Methylindole-2-carboxylic acid

98%

Synonym(s):

NSC 68357

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

212-213 °C (dec.) (lit.)

SMILES string

Cn1c(cc2ccccc12)C(O)=O

InChI

1S/C10H9NO2/c1-11-8-5-3-2-4-7(8)6-9(11)10(12)13/h2-6H,1H3,(H,12,13)

InChI key

MAHAMBLNIDMREX-UHFFFAOYSA-N

General description

1-Methylindole-2-carboxylic acid reacts with thionyl chloride to yield sulfinyl chlorides.

Application

  • Reactant for preparation of keto-indoles as novel indoleamine 2,3-dioxygenase (IDO) inhibitors
  • Reactant for synthesis of fenbufen and ethacrynic acid derivatives as potential antitumor agents via amide coupling reactions
  • Reactant for diastereoselective synthesis of vinylated heterocycles via ruthenium-catalyzed oxidative vinylation with alkenes
  • Reactant for synthesis of 2,3-dihalo indoles via hypervalent iodine mediated decarboxylative halogenation
  • Reactant for preparation of α-ketoamides as cathepsin S inhibitors with potential applications against tumor invasion and angiogenesis
  • Reactant for preparation of anthranilic acid mimics as bacterial translation inhibitors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of Indole Derivatives with Thionyl and Sulfuryl Chlorides.
Szmuszkovicz J.
The Journal of Organic Chemistry, 29(1), 178-184 (1964)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service