Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

134139

Sigma-Aldrich

5-Methylindole-3-carboxaldehyde

97%

Synonym(s):

NSC 88872

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO
CAS Number:
Molecular Weight:
159.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

148-151 °C (lit.)

functional group

aldehyde

SMILES string

Cc1ccc2[nH]cc(C=O)c2c1

InChI

1S/C10H9NO/c1-7-2-3-10-9(4-7)8(6-12)5-11-10/h2-6,11H,1H3

InChI key

ZTNQWTPWKNDRNF-UHFFFAOYSA-N

Application

5-Methylindole-3-carboxaldehyde was used in the synthesis of novel flexible tripodal ligand derived from 3-methylindole and its mononuclear Zn(II),Cu(II), Ni(II), Hg(II) and Pd(II) complexes[1].
Reactant for preparation of:
  • Tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
  • Antimicrobial agents
  • Anti-proliferative agents
  • Antiandrogens effective against multiple clinically relevant androgen receptor mutants
  • Anticancer agents
  • Inhibitors of Mycobacterium tuberculosis protein tyrosine phosphatase B
  • Bovine viral diarrhea virus (BVDV) inhibitors
  • Potent thermal sensitizing agents
  • Analogues of marine alkaloid nortopsentin as anticancer agents
  • β3-adrenergic receptor agonists

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new tripodal poly-imine indole-containing ligand: Synthesis, complexation, spectroscopic and theoretical studies.
Pedras B, et al.
Inorgorganica Chimica Acta, 362(8), 2627-2635 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service