Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

133655

Sigma-Aldrich

4′-Chloroacetoacetanilide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CONHC6H4Cl
CAS Number:
Molecular Weight:
211.64
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

131-134 °C (lit.)

solubility

acetone: soluble 25 mg/mL, clear, colorless to light yellow

SMILES string

CC(=O)CC(=O)Nc1ccc(Cl)cc1

InChI

1S/C10H10ClNO2/c1-7(13)6-10(14)12-9-4-2-8(11)3-5-9/h2-5H,6H2,1H3,(H,12,14)

InChI key

JMRJWEJJUKUBEA-UHFFFAOYSA-N

General description

4′-Chloroacetoacetanilide was trilithiated with lithium diisopropylamide and condensed with several aromatic esters, followed by neutralization, separate acid cyclization and rearrangements to yield 4-anilino-6-aryl-2H-pyran-2-ones. It undergoes three-component reaction with a mixture of aromatic aldehyde and 5-aminotetrazole to form N,7-diaryl-5-methyl-4,7-dihydrotetrazolo[1,5-a]pyrimidine-6-carboxamide.

Application

4′-Chloroacetoacetanilide was used in recombinant androgen receptor competitive binding assay for analysis of natural, synthetic and environmental chemicals.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and antimicrobial activity of n, 7-diaryl-5-methyl-4, 7-dihydrotetrazolo [1, 5-a] pyrimidine-6-arboxamides.
Gein VL, et al.
Pharmaceutical Chemistry Journal, 44(7), 366-369 (2010)
Preparation of 4-anilino-6-aryl-2 H-pyran-2-ones from trilithiated acetoacetanilides and aromatic esters.
Downs JR, et al.
Canadian Journal of Chemistry, 82(5), 659-664 (2004)
Hong Fang et al.
Chemical research in toxicology, 16(10), 1338-1358 (2003-10-21)
A number of environmental and industrial chemicals are reported to possess androgenic or antiandrogenic activities. These androgenic endocrine disrupting chemicals may disrupt the endocrine system of humans and wildlife by mimicking or antagonizing the functions of natural hormones. The present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service