Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

133310

Sigma-Aldrich

Aniline hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NH2 · HCl
CAS Number:
Molecular Weight:
129.59
Beilstein:
3593823
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

4.46 (vs air)

Assay

97%

mp

196-198 °C (lit.)

SMILES string

Cl.Nc1ccccc1

InChI

1S/C6H7N.ClH/c7-6-4-2-1-3-5-6;/h1-5H,7H2;1H

InChI key

MMCPOSDMTGQNKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Aniline hydrochloride was used in the preparation of polyaniline coated poly(styrene-co-styrene sulfonate) nanoparticles. It was used to study the induction of Nei-like DNA glycosylases (NEIL1/2)-mediated base excision repair(BER) in rat spleen and 8-oxoguanine glycosylase 1-mediated BER due to aniline exposure.

Biochem/physiol Actions

The acute toxicity of aniline involves its activation in vivo to 4-hydroxyaniline and the formation of adducts with hemoglobin. In erythrocytes, this is associated with the release of iron and the accumulation of methemoglobin and the development of hemolytic anemia and inflammation of the spleen. Tumor formation is often observed in the spleen on prolonged administration.

Other Notes

This product has been replaced by A8524-ALDRICH | Aniline hydrochloride ≥99%

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

381.2 °F - closed cup

Flash Point(C)

194 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of PANI-coated poly (styrene-co-styrene sulfonate) nanoparticles.
Kim BJ, et al.
Polymer, 43(1), 111-116 (2002)
Huaxian Ma et al.
Toxicology and applied pharmacology, 251(1), 1-7 (2010-12-15)
The mechanisms by which aniline exposure elicits splenotoxic response, especially the tumorigenic response, are not well-understood. Earlier, we have shown that aniline-induced oxidative stress is associated with increased oxidative DNA damage in rat spleen. The base excision repair (BER) pathway
Timo Stahl et al.
Journal of the American Chemical Society, 135(4), 1248-1251 (2013-01-15)
Heterolytic splitting of the Si-H bond mediated by a Ru-S bond forms a sulfur-stabilized silicon cation that is sufficiently electrophilic to abstract fluoride from CF(3) groups attached to selected anilines. The ability of the Ru-H complex, generated in the cooperative
Dongdong Liang et al.
Chemical communications (Cambridge, England), 49(2), 173-175 (2012-11-22)
An efficient synthesis of free (NH)-phenanthridinones through Pd-catalyzed C(sp(2))-H aminocarbonylation of unprotected o-arylanilines under an atmospheric pressure of CO has been developed. Some ortho heteroarene substituted anilines as well as N-alkyl protected o-arylanilines are also suitable substrates for this C-H
Roman Spesyvtsev et al.
Faraday discussions, 157, 165-179 (2012-12-13)
Femtosecond time-resolved photoelectron imaging is employed to investigate ultrafast electronic relaxation in aniline, a prototypical aromatic amine. The molecule is excited at wavelengths between 269 and 238 nm. We observe that the S2(pi3s/pi sigma*) state is populated directly during the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service