Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

130982

Sigma-Aldrich

4,6-Dichlororesorcinol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(OH)2
CAS Number:
Molecular Weight:
179.00
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

254 °C (lit.)

mp

104-106 °C (lit.)

functional group

chloro

SMILES string

Oc1cc(O)c(Cl)cc1Cl

InChI

1S/C6H4Cl2O2/c7-3-1-4(8)6(10)2-5(3)9/h1-2,9-10H

InChI key

GRLQBYQELUWBIO-UHFFFAOYSA-N

Application

4,6-Dichlororesorcinol was used as an exotemplate to self-organize two aza-heterocyclic units by OH−N hydrogen bonds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrada Stefania Trita et al.
Organic letters, 15(14), 3798-3801 (2013-07-09)
The construction of pyridine-containing macrocyclic architectures using a nonmetallic template is described. 4,6-Dichlororesorcinol was used as an exotemplate to self-organize two aza-heterocyclic units by OH···N hydrogen bonds. Subsequent sequential double olefin metathesis/hydrogenation reactions employing a single ruthenium-alkylidene precatalyst open access

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service