Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

130788

Sigma-Aldrich

Uracil

98%

Synonym(s):

2,4(1H,3H)-Pyrimidinedione, 2,4-Dihydroxypyrimidine, 2,4-Pyrimidinediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2O2
CAS Number:
Molecular Weight:
112.09
Beilstein:
507828
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

mp

>300 °C (lit.)

SMILES string

O=C1NC=CC(=O)N1

InChI

1S/C4H4N2O2/c7-3-1-2-5-4(8)6-3/h1-2H,(H2,5,6,7,8)

InChI key

ISAKRJDGNUQOIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra M Carvalho et al.
PloS one, 8(3), e58492-e58492 (2013-03-19)
Links between carbohydrate metabolism and virulence in Streptococcus pneumoniae have been recurrently established. To investigate these links further we developed a chemically defined medium (CDM) and standardized growth conditions that allowed for high growth yields of the related pneumococcal strains
Daisuke Katagiri et al.
Journal of the American Society of Nephrology : JASN, 24(12), 2034-2043 (2013-10-05)
Accumulating evidence of the beyond-glucose lowering effects of a gut-released hormone, glucagon-like peptide-1 (GLP-1), has been reported in the context of remote organ connections of the cardiovascular system. Specifically, GLP-1 appears to prevent apoptosis, and inhibition of dipeptidyl peptidase-4 (DPP-4)
Torkild Visnes et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 364(1517), 563-568 (2008-11-15)
Uracil in DNA may result from incorporation of dUMP during replication and from spontaneous or enzymatic deamination of cytosine, resulting in U:A pairs or U:G mismatches, respectively. Uracil generated by activation-induced cytosine deaminase (AID) in B cells is a normal
Jaunius Urbonavicius et al.
Methods in enzymology, 425, 103-119 (2007-08-04)
Formation of 5-methyluridine (ribothymidine) at position 54 of the T-psi loop of tRNA is catalyzed by site-specific tRNA methyltransferases (tRNA[uracil-54,C5]-MTases). In eukaryotes and many bacteria, the methyl donor for this reaction is generally S-adenosyl-L-methionine (S-AdoMet). However, in other bacteria, like
Pietro Andreone et al.
Gastroenterology, 147(2), 359-365 (2014-05-14)
The interferon-free regimen of ABT-450 (a protease inhibitor), ritonavir, ombitasvir (an NS5A inhibitor), dasabuvir (a non-nucleoside polymerase inhibitor), and ribavirin has shown efficacy in patients with hepatitis C virus (HCV) genotype 1b infection-the most prevalent subgenotype worldwide. We evaluated whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service