Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

130435

Sigma-Aldrich

4-Dimethylamino-2-methylazobenzene

Synonym(s):

N,N-Dimethyl-4-phenylazo-m-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N=NC6H3(CH3)N(CH3)2
CAS Number:
Molecular Weight:
239.32
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

mp

67-69 °C (lit.)

SMILES string

CN(C)c1ccc(\N=N\c2ccccc2)c(C)c1

InChI

1S/C15H17N3/c1-12-11-14(18(2)3)9-10-15(12)17-16-13-7-5-4-6-8-13/h4-11H,1-3H3/b17-16+

InChI key

QXTOLYZIJUDOIT-WUKNDPDISA-N

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Yanagi et al.
Journal of the National Cancer Institute, 73(4), 887-894 (1984-10-01)
A probably promoter-specific decrease of L-type pyruvate kinase (L-PK) in Wistar rat liver is described. The possibility of utilizing the decrease in L-PK activity for screening of hepatic promoters is discussed. A significantly decreased level of activity of L-PK was
A R Samuels et al.
Biochimica et biophysica acta, 759(1-2), 23-31 (1983-08-23)
Following intravenous administration to rats of the azo dye hepatocarcinogen 3'-methyl-N,N-dimethyl-4-aminoazo-[14C]benzene, 60-70% of the injected dose was recovered in bile in 2 h. Approximately 10% of bile radioactivity was trichloroacetic acid-precipitable, not extracted by n-butanol and non-dialyzable. Sodium dodecyl sulfate-polyacrylamide
Inhibition of mitochondrial electron transport and energy transduction by 2-methyl-4-dimethylaminoazobenzene in vitro.
P Saikumar et al.
Indian journal of biochemistry & biophysics, 21(5), 309-313 (1984-10-01)
Early events during liver carcinogenesis involving two carcinogen:protein complexes.
G R Blackburn et al.
Cancer research, 41(10), 4039-4049 (1981-10-01)
R Daoust
Chemico-biological interactions, 48(2), 221-235 (1984-02-01)
In order to obtain a more precise definition of the conditions under which 2-methyl-4-dimethylaminoazobenzene (2-Me-DAB) and liver cell proliferation play a role in the initiation of hepatocarcinogenesis, the toxicity of 2-Me-DAB for normal and partially hepatectomized rats was investigated. Continuous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service