Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

130192

Sigma-Aldrich

trans-3-Nitrocinnamic acid

97%

Synonym(s):

trans-3-(3-Nitrophenyl)acrylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
193.16
Beilstein:
2211201
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

200-202 °C (lit.)

functional group

carboxylic acid
nitro

SMILES string

OC(=O)\C=C\c1cccc(c1)[N+]([O-])=O

InChI

1S/C9H7NO4/c11-9(12)5-4-7-2-1-3-8(6-7)10(13)14/h1-6H,(H,11,12)/b5-4+

InChI key

WWXMVRYHLZMQIG-SNAWJCMRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans-3-Nitrocinnamic acid was used in the preparation of cyclobutane adduct[1].

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unusual [2+ 2] photocycloaddition between tryptamine and 3-nitrocinnamic acid in the solid state.
Ito Y, et al.
Chemistry Letters (Jpn), 29(3), 288-289 (2000)
K Oztürk et al.
Toxicology letters, 108(1), 63-68 (1999-09-03)
Four nitroarenes were tested in two standard genotoxicity assay systems using three well-known bacterial tester strains. The results were as follows: 4-nitroquinoline l-oxide (4NQO) was positive in Quillardet and Hofnung's SOS chromotest using Escherichia coli strain PQ37 both in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service