Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

129739

Sigma-Aldrich

3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate

97%

Synonym(s):

MBTH hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9N3S · HCl · xH2O
CAS Number:
Molecular Weight:
215.70 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

276-278 °C (dec.) (lit.)

SMILES string

Cl[H].[H]O[H].CN1\C(Sc2ccccc12)=N\N

InChI

1S/C8H9N3S.ClH.H2O/c1-11-6-4-2-3-5-7(6)12-8(11)10-9;;/h2-5H,9H2,1H3;1H;1H2/b10-8-;;

InChI key

IYXXQOGEFHAQGU-GPWRMYTLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate (MBTH hydrochloride hydrate) was used as an electrophilic coupling reagent for the determination of residual chlorine[1].
Chromogenic reagent for the determination of benzodiazepines,[2] cholesterol,[3] and enzyme activity.[4] Used in the synthesis of benzothiazolium azo dyes.[5]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bull. Soc. Chim. Belg., 102, 233-233 (1993)
J N Rodríguez-López et al.
Analytical biochemistry, 216(1), 205-212 (1994-01-01)
A continuous spectrophotometric method for the rapid determination of monophenolase activity of tyrosinase is described. This method is based on the coupling reaction between 3-methyl-2-benzothiazolinone hydrazone (MBTH) and the quinone products of the oxidation of various monophenols in the presence
Dyes and Pigments, 21, 159-159 (1993)
Indian J. Chem. B, 32, 44-44 (1993)
Riyad Ahmed Al-Okab et al.
Journal of hazardous materials, 170(1), 292-297 (2009-05-30)
Novel, sensitive and rapid spectrophotometric methods, using phenoxazine (PNZ), 2-chlorophe-noxazine (CPN) and 2-trifluoromethylphenoxazine (TPN) as chromogenic reagents for the determination of residual chlorine are proposed. The methods are based on the reduction of chlorine by an electrophilic coupling reagent, 3-methyl-2-benzothiazoline

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service