Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12973

Sigma-Aldrich

Benzoyl chloride polymer bound

100-300 mesh, extent of labeling: ~1.5 mmol/g loading

Synonym(s):

(Chlorocarbonyl)polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: amine reactive

extent of labeling

~1.5 mmol/g loading

matrix

crosslinked with 2% DVB

particle size

100-300 mesh

Other Notes

Support for SPOS of polyheterocyclic isoxazoles; Scavenger for removing excess amines

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E.J. Kantorowski et al.
The Journal of Organic Chemistry, 62, 6797-6797 (1997)
Jürgen Eich et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 37(1), 84-93 (2019-11-08)
The described methods are able to analyse the triphenylmethane dyes malachite green (MG), crystal violet (CV) and brilliant green (BG) as well as their leuco metabolites leuco malachite green (LMG), leuco crystal violet (LCV) and leuco brilliant green (LBG) on the
S.W. Kaldor et al.
Tetrahedron Letters, 37, 7193-7193 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service