Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

129127

Sigma-Aldrich

1-Chloro-3,5-dibromobenzene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2C6H3Cl
CAS Number:
Molecular Weight:
270.35
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

91-94 °C (lit.)

functional group

bromo
chloro

SMILES string

Clc1cc(Br)cc(Br)c1

InChI

1S/C6H3Br2Cl/c7-4-1-5(8)3-6(9)2-4/h1-3H

InChI key

FNKCOUREFBNNHG-UHFFFAOYSA-N

Application

1-Chloro-3,5-dibromobenzene was used in the preparation of aryl bromide by the process of deprotonation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffery A Gladding et al.
Tetrahedron, 67(35), 6697-6706 (2011-10-25)
Studies directed towards the synthesis of the architecturally complex marine natural product sporolide B are described. Synthetic analysis suggested advanced hydroquinone and benzodiquinane fragments, which upon elaboration were successfully united via an ester linkage. Macrocyclization studies were then carried out

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service