Skip to Content
MilliporeSigma
All Photos(1)

Documents

128325

Sigma-Aldrich

2-(N-Ethyl-N-m-toluidino)ethanol

98%

Synonym(s):

N-Ethyl-N-(2-hydroxyethyl)-m-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4N(C2H5)CH2CH2OH
CAS Number:
Molecular Weight:
179.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.555 (lit.)

bp

114-115 °C/1 mmHg (lit.)

density

1.019 g/mL at 25 °C (lit.)

SMILES string

CCN(CCO)c1cccc(C)c1

InChI

1S/C11H17NO/c1-3-12(7-8-13)11-6-4-5-10(2)9-11/h4-6,9,13H,3,7-8H2,1-2H3

InChI key

KRNUKKZDGDAWBF-UHFFFAOYSA-N

Application

2-(N-Ethyl-N-m-toluidino)ethanol may be used in chemical synthesis studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

298.4 °F - DIN 51758

Flash Point(C)

148 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service