Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

127159

Sigma-Aldrich

N-(2-Aminoethyl)-1,3-propanediamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)3NHCH2CH2NH2
CAS Number:
Molecular Weight:
117.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.4815 (lit.)

density

0.928 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

NCCCNCCN

InChI

1S/C5H15N3/c6-2-1-4-8-5-3-7/h8H,1-7H2

InChI key

DTSDBGVDESRKKD-UHFFFAOYSA-N

Application

N-(2-Aminoethyl)-1,3-propanediamine has been used as a templating agent in the synthesis of a new open framework iron(III) phosphite {(C5H18N3)[Fe3(HPO3)6].3H2O}. It has also been used in the preparation of dinitrocobalt(III) compound: (11-amino-4-methyl-5,8-diazaundeca-2,4-dien-2-olato-kappa4N(5,8,11),O)-dinitrocobalt(III), [Co(C10H20N3O)(NO2)2].

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Chun et al.
Acta crystallographica. Section C, Crystal structure communications, 57(Pt 1), 33-35 (2001-02-15)
The reaction of Co(acac)3 with N-(2-aminoethyl)-1,3-propanediamine in the presence of NaNO2 results in the preparation of an unexpected dinitrocobalt(III) compound, (11-amino-4-methyl-5,8-diazaundeca-2,4-dien-2-olato-kappa4N(5,8,11),O)-dinitrocobalt(III), [Co(C10H20N3O)(NO2)2], containing the tetradentate anion of 11-amino-4-methyl-5,8-diazaundeca-2,4-dien-2-ol. Two isomers of the compound were obtained by recrystallization of the crude
U-Chan Chung et al.
Inorganic chemistry, 45(22), 8965-8972 (2006-10-24)
A new open framework iron(III) phosphite with formula (C5H18N3)[Fe3(HPO3)6].3H2O has been prepared by hydrothermal synthesis with N-(2-aminoethyl)-1,3-propanediamine as a templating agent. The crystal structure was solved from single-crystal X-ray diffraction data in the trigonal space group R. The unit cell
Naciye Esma Tirtom et al.
Physical chemistry chemical physics : PCCP, 22(21), 11928-11935 (2020-05-21)
Polyamines are naturally occurring cationic molecules in cells. In addition to their roles in modulating gene expression and cell proliferation, they have been shown to stimulate DNA recombination. The molecular mechanism for stimulation is not clear. We utilized single-molecule tethered
Lucas Vu et al.
Theranostics, 2(12), 1160-1173 (2013-02-06)
A focused library of twenty-one cationic poly(amino ethers) was synthesized following ring-opening polymerization of two diglycidyl ethers by different oligoamines. The polymers were screened in parallel for plasmid DNA (pDNA) delivery, and transgene expression efficacies of individual polymers were compared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service