Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

126926

Sigma-Aldrich

N,N,N′,N′-Tetramethyl-1,3-propanediamine

98%

Synonym(s):

1,3-Bis(dimethylamino)propane, TMPDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2N(CH2)3N(CH3)2
CAS Number:
Molecular Weight:
130.23
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor pressure

760 mmHg ( 145 °C)

Assay

98%

refractive index

n20/D 1.4234 (lit.)

bp

145-146 °C (lit.)

density

0.779 g/mL at 25 °C (lit.)

SMILES string

CN(C)CCCN(C)C

InChI

1S/C7H18N2/c1-8(2)6-5-7-9(3)4/h5-7H2,1-4H3

InChI key

DMQSHEKGGUOYJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service