Skip to Content
MilliporeSigma
All Photos(1)

Documents

126330

Sigma-Aldrich

2,3-Dimethoxyphenol

98%

Synonym(s):

Pyrogallol 1,2-dimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3OH
CAS Number:
Molecular Weight:
154.16
Beilstein:
1910045
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.539 (lit.)

bp

233-234 °C (lit.)

density

1.182 g/mL at 25 °C (lit.)

SMILES string

COc1cccc(O)c1OC

InChI

1S/C8H10O3/c1-10-7-5-3-4-6(9)8(7)11-2/h3-5,9H,1-2H3

InChI key

QSZCGGBDNYTQHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Dimethoxyphenol is a plant-derived phenylpropanoid compound that is glycosidated to form glycoside compound.

Application

2,3-Dimethoxyphenol has been used to study the nitrosative deamination of DNA bases induced by reactive nitrogen species (RNS) while studying the cause of mutagenesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shashi Kant Bhatia et al.
Bioresource technology, 289, 121704-121704 (2019-07-06)
Rhodococcus sp. YHY01 was studied to utilize various lignin derived aromatic compounds. It was able to utilize p-coumaric acid, cresol, and 2,6 dimethoxyphenol and resulted in biomass production i.e. 0.38 g dcw/L, 0.25 g dcw/L and 0.1 g dcw/L, and lipid accumulation i.e.
K Rybczyńska-Tkaczyk et al.
Ecotoxicology and environmental safety, 191, 110203-110203 (2020-01-24)
The aim of this study was to evaluate of possibility of biotransformation and toxicity effect of monoanthraquinone dyes in cultures of Bjerkandera adusta CCBAS 930. Phenolic compounds, free radicals, phytotoxicity (Lepidium sativum L.), ecotoxicity (Vibrio fischeri) and cytotoxicity effect were
Qi Luo et al.
Environmental science & technology, 52(18), 10617-10626 (2018-08-28)
Perfluorooctanesulfonate (PFOS) is a compound that has wide applications with extreme persistence in the environment and the potential to bioaccumulate, and could induce adverse effects to ecosystems. We investigated the degradation of PFOS by laccase-induced enzyme catalyzed oxidative humification reactions
Márcia Pessêgo et al.
Journal of pharmacy & bioallied sciences, 3(1), 128-134 (2011-03-25)
Nitrosative deamination of DNA bases induced by reaction with reactive nitrogen species (RNS) has been pointed out as a probable cause of mutagenesis. (Poly)phenols, present in many food items from the Mediterranean diet, are believed to possess antinitrosating properties due
O Saoudi et al.
International journal of biological macromolecules, 128, 681-691 (2019-02-04)
This investigation may be of interest for researchers working on the determination of several biocatalytic properties of the laccase from Trametes versicolor. So, We will treated the effects of pH, temperature, several organic components and heavy metals by performing enzyme

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service