Skip to Content
MilliporeSigma
All Photos(1)

Documents

126012

Sigma-Aldrich

2,6-Dichlorophenylacetonitrile

97%

Synonym(s):

2,6-Dichlorobenzyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CH2CN
CAS Number:
Molecular Weight:
186.04
Beilstein:
1910482
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

73-75 °C (lit.)

SMILES string

Clc1cccc(Cl)c1CC#N

InChI

1S/C8H5Cl2N/c9-7-2-1-3-8(10)6(7)4-5-11/h1-3H,4H2

InChI key

AOEJUUCUKRUCEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Dichlorophenylacetonitrile was found to degrade rapidly in non-sterile soils but not in azide treated soils.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrolysis of 2,6-dichlorobenzonitrile in soils.
G G Briggs et al.
Journal of agricultural and food chemistry, 18(1), 97-99 (1970-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service