Skip to Content
MilliporeSigma
All Photos(1)

Documents

125202

Sigma-Aldrich

3-Chloropropionamide

98%

Synonym(s):

beta-Chloropropionamide, chloropropionamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2CONH2
CAS Number:
Molecular Weight:
107.54
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

98-101 °C (lit.)

SMILES string

NC(=O)CCCl

InChI

1S/C3H6ClNO/c4-2-1-3(5)6/h1-2H2,(H2,5,6)

InChI key

JQDXZJYAUSVHDH-UHFFFAOYSA-N

Application

3-Chloropropionamide was used as alkylating agent in the synthesis of 1-(2-carbamoyl-ethyl)indole-3-acetic acid.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of N-alkyl substituted bioactive indolocarbazoles related to Go6976.
Roy S, et al.
Tetrahedron, 62(33), 7838-7845 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service