Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

124257

Sigma-Aldrich

N-Butyldiethanolamine

98%

Synonym(s):

2,2′-Butyliminodiethanol, N,N-Bis(2-hydroxyethyl)butylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3N(CH2CH2OH)2
CAS Number:
Molecular Weight:
161.24
Beilstein:
1739642
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

vapor density

5.5 (vs air)

vapor pressure

1 mmHg ( 25 °C)

Assay

98%

refractive index

n20/D 1.463 (lit.)

bp

273-275 °C/741 mmHg (lit.)

mp

−70 °C (lit.)

density

0.986 g/mL at 25 °C (lit.)

SMILES string

CCCCN(CCO)CCO

InChI

1S/C8H19NO2/c1-2-3-4-9(5-7-10)6-8-11/h10-11H,2-8H2,1H3

InChI key

GVNHOISKXMSMPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Butyldiethanolamine is a tertiary amine. Densities and excess molar volumes of its binary mixtures with water have been evaluated in the temperature range of 298.15 to 353.15K.

Application

N-Butyldiethanolamine (H2bdea) may be used in the preparation of following copper(II) compounds [Hdnba = 3,5-dinitrobenzoic acid, Hpta = p-toluic acid, H2tpa = terephthalic acid):
  • mononuclear [Cu(Hbdea)2]·2Hdnba
  • dinuclear [Cu2(μ-Hbdea)2(N3)2]
  • [Cu2(μ-Hbdea)2(pta)2]·2H2O
  • 1D polymeric [Cu2(μ-Hbdea)2(μ-tpa)]n.2nH2O

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

265.1 °F - closed cup

Flash Point(C)

129.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Densities and volumetric properties of the aqueous solutions of 2-amino-2-methyl-1-propanol, n-butyldiethanolamine and n-propylethanolamine at temperatures from 298.15 to 353.15 K.
Chan C, et al.
Fluid Phase Equilibria, 198(2), 239-250 (2002)
Katrin R Gruenwald et al.
Dalton transactions (Cambridge, England : 2003), (12)(12), 2109-2120 (2009-03-11)
The new mononuclear [Cu(Hbdea)(2)].2Hdnba (), dinuclear [Cu(2)(mu-Hbdea)(2)(N(3))(2)] () and [Cu(2)(mu-Hbdea)(2)(pta)(2)].2H(2)O (), and 1D polymeric [Cu(2)(mu-Hbdea)(2)(mu-tpa)](n).2nH(2)O () copper(II) compounds have been prepared by self-assembly, in aqueous alkali medium and at ambient conditions, from Cu(II) acetate, N-butyldiethanolamine (H(2)bdea) and the corresponding auxiliary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service