11998
N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride
Sivento quality, 45-55% in methanol
About This Item
Recommended Products
grade
Sivento quality
manufacturer/tradename
Sivento Chemie GmbH
concentration
45-55% in methanol
refractive index
n20/D 1.397 (lit.)
density
0.93 g/mL at 20 °C (lit.)
InChI
1S/C9H24NO3Si.ClH/c1-10(2,3)8-7-9-14(11-4,12-5)13-6;/h7-9H2,1-6H3;1H/q+1;/p-1
InChI key
FYZFRYWTMMVDLR-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3
Target Organs
Eyes,Central nervous system, Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
51.8 °F - closed cup
Flash Point(C)
11 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service