Skip to Content
MilliporeSigma
All Photos(1)

Documents

118710

Sigma-Aldrich

2-Methyl-1,4-pentadiene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2C(CH3)=CH2
CAS Number:
Molecular Weight:
82.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.406 (lit.)

bp

56 °C (lit.)

density

0.692 g/mL at 25 °C (lit.)

SMILES string

CC(=C)CC=C

InChI

1S/C6H10/c1-4-5-6(2)3/h4H,1-2,5H2,3H3

InChI key

DRWYRROCDFQZQF-UHFFFAOYSA-N

General description

2-Methyl-1,4-pentadiene forms an organotrifluoroborate reagent by cross-metathesis reaction with vinylboronate. It reacts with bromo or iodo benzene in the presence of piperidine and palladium acetate-tri-o-tolylphosphine catalyst to give 1-phenyl-5-piperidino-3-alkenes.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

28.4 °F - closed cup

Flash Point(C)

-2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jorge Esteban et al.
Organic letters, 10(21), 4843-4846 (2008-10-08)
The key C10-C26 fragment in a total synthesis of (-)-amphidinolide E has been prepared from an oxolane-containing C10-C17 segment (9, derived from L-glutamic acid) via a Julia-Kocienski reaction with aldehyde 3, followed by a Sharpless AD to obtain the desired
Palladium-catalyzed arylation and vinylation of 1, 4-dienes.
Bender DD, et al.
The Journal of Organic Chemistry, 47(7), 1278-1284 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service