Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

118486

Sigma-Aldrich

2,3-Dimethyl-3-pentanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(CH3)(OH)CH(CH3)2
CAS Number:
Molecular Weight:
116.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.428 (lit.)

bp

140 °C (lit.)

density

0.833 g/mL at 25 °C (lit.)

SMILES string

CCC(C)(O)C(C)C

InChI

1S/C7H16O/c1-5-7(4,8)6(2)3/h6,8H,5H2,1-4H3

InChI key

RFZHJHSNHYIRNE-UHFFFAOYSA-N

Application

2,3-Dimethyl-3-pentanol was used in a study to determine the uptake rates for different compounds of various polarities by polydimethylsiloxane-based permeation passive air sampler.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suresh Seethapathy et al.
Journal of chromatography. A, 1217(50), 7907-7913 (2010-11-12)
Polydimethylsiloxane (PDMS) has low permeability towards water vapour and low energy of activation of permeation towards volatile organic compounds (VOCs) when compared to many other polymers. Suitability of the material for use in permeation-type passive air samplers was tested as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service