Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

116335

Sigma-Aldrich

2-Chloro-6-methylpyridine

99%

Synonym(s):

6-Chloro-2-picoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6ClN
CAS Number:
Molecular Weight:
127.57
Beilstein:
107187
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.527 (lit.)

bp

64-68 °C/10 mmHg (lit.)

density

1.167 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Cc1cccc(Cl)n1

InChI

1S/C6H6ClN/c1-5-3-2-4-6(7)8-5/h2-4H,1H3

InChI key

GXZDYRYYNXYPMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloro-6-methylpyridine has been used for experimental FTIR and FT-Raman spectroscopic analyses, as well as for conformational studies[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Arjunan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 92, 305-317 (2012-03-27)
Experimental FTIR and FT-Raman spectroscopic analysis of 2-chloro-4-methyl-3-nitropyridine (2C4M3NP) and 2-chloro-6-methylpyridine (2C6MP) have been performed. A detailed quantum chemical calculations have been carried out using B3LYP and B3PW91 methods with 6-311++G** and cc-pVTZ basis sets. Conformation analysis was carried for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service