Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11558

Sigma-Aldrich

Lithium L-aziridine-2-carboxylate

≥97.0% (dried material, NT)

Synonym(s):

L-Aziridine-2-carboxylic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4LiNO2
CAS Number:
Molecular Weight:
93.01
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥97.0% (dried material, NT)

optical activity

[α]20/D −42±2°, c = 1% in H2O

impurities

~3% water

application(s)

peptide synthesis

SMILES string

[Li+].[O-]C(=O)[C@@H]1CN1

InChI

1S/C3H5NO2.Li/c5-3(6)2-1-4-2;/h2,4H,1H2,(H,5,6);/q;+1/p-1/t2-;/m0./s1

InChI key

ZGQWDLXRNCMIHH-DKWTVANSSA-M

Application

Lithium L-aziridine-2-carboxylate may be used as a chelate ligand to form complexes with transition metal salts.[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Chiral building block; protected derivatives are used for the synthesis of various amino acids by ring-opening with nucleophiles [2][3][4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Nakajima et al.
Bulletin of the Chemical Society of Japan, 56, 520-520 (1983)
Y. Kogami et al.
Bulletin of the Chemical Society of Japan, 60, 2963-2963 (1987)
R.J. Parry et al.
Journal of the American Chemical Society, 107, 2512-2512 (1985)
Transition metal complexes of l-aziridine-carboxylate.
Hauck T, et al.
Inorgorganica Chimica Acta, 235(1-2), 391-396 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service